Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Phthalide 99%

Phthalide 99%

Plastic bottle, 500 g
Catalog # ACRO131095000
Irritant
Phthalide 99%
Catalog # ACRO131095000
CAS Number:  87-41-2
Phthalide 99%
Phthalide 99%

Plastic bottle, 500 g

CAS Number:  87-41-2
Price
£88.60
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Isobenzofuran-1(3H)-one
  • CAS Number:87-41-2
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00005906
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:70…73 °C
  • Density:1,1 g/cm³ (30 °C)
  • Flash Pt:152 °C
  • EINECS:201-744-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 73 °C
  • GC:
    ≥98.5 %
  • UV:
    at 230 nm A: ≤0.15
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.001 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications