L(-)-Menthol 99.5%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
- CAS Number:2216-51-5
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00062979
- Molecular Weight:156.27 g/mol
- Merck Index:13,05861
- Boiling Pt:212 °C (760 mmHg)
- Melting Pt:41…43 °C
- Density:0.89 g/cm³
- Flash Pt:94 °C
- Storage Temperature:Ambient
- EINECS:218-690-9
Documents
Specifications
Specification Test Results
- Appearance:Colorless to white Crystals or crystalline needles
- Infrared spectrum:Conforms
- GC:≥99.4 %
- Specific optical rotation:-48° to -51° (20°C, 589 nm) (c=10, EtOH)
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



