Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Fluorophenol 98%

2-Fluorophenol 98%

Glass bottle for solids, 100 g
Catalog # ACRO119541000
Irritant
Flammable
2-Fluorophenol 98%
Catalog # ACRO119541000
CAS Number:  367-12-4
2-Fluorophenol 98%
2-Fluorophenol 98%

Glass bottle for solids, 100 g

CAS Number:  367-12-4
Price
£126.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  o-Fluorophenol
  • CAS Number:367-12-4
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002155
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:152 °C (760 mmHg)
  • Melting Pt:16.1 °C
  • Density:1.256 g/cm³
  • Flash Pt:46 °C
  • Storage Temperature:Ambient
  • EINECS:206-681-2
  • UN:1993
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5134 to 1.5154 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • UV:
    at 230 nm A: ≤0.2
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤2 ppm
  • Acidity:
    ≤0.0003 meq/g
  • Filtered to 0.2 micrometer:
    Filtered

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications