2-Fluorophenol 98%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: o-Fluorophenol
- CAS Number:367-12-4
- Formula:C₆H₅FO
- MDL Number:MFCD00002155
- Molecular Weight:112.1 g/mol
- Boiling Pt:152 °C (760 mmHg)
- Melting Pt:16.1 °C
- Density:1.256 g/cm³
- Flash Pt:46 °C
- Storage Temperature:Ambient
- EINECS:206-681-2
- UN:1993
- ADR:3,III
Documents
ACRO11954
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5134 to 1.5154 (20°C, 589 nm)
- GC:≥97.5 %
- UV:at 230 nm A: ≤0.2
- Color scale:≤10 APHA
- Residue after evaporation:≤2 ppm
- Acidity:≤0.0003 meq/g
- Filtered to 0.2 micrometer:Filtered
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



