9-Fluorenone 99+%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C (760 mmHg)
- Melting Pt:81…85 °C
- Density:0,9 g/cm³ (20 °C)
- Flash Pt:163 °C
- Storage Temperature:Ambient
- EINECS:207-630-7
- UN:3077
Documents
ACRO11920
SDS
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:81 to 85 °C
- GC:≥99.0 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



