9-Fluorenone 99+%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 9H-Fluoren-9-one
- CAS Number:486-25-9
- Formula:C₁₃H₈O
- MDL Number:MFCD00001141
- Molecular Weight:180.21 g/mol
- Boiling Pt:342 °C (760 mmHg)
- Melting Pt:81…85 °C
- Density:0,9 g/cm³ (20 °C)
- Flash Pt:163 °C
- Storage Temperature:Ambient
- EINECS:207-630-7
- UN:3077
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:81 to 85 °C
- GC:≥99.0 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Plastic bottle
- Pk:100 g



