2,6-Xylidine 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline
- CAS Number:87-62-7
- Formula:C₈H₁₁N
- MDL Number:MFCD00007747
- Molecular Weight:121.18 g/mol
- Merck Index:12,10217
- Boiling Pt:216 °C (760 mmHg)
- Melting Pt:10…12 °C
- Density:0.98 g/cm³
- Flash Pt:91 °C
- Storage Temperature:Ambient
- EINECS:201-758-7
- UN:1711
- ADR:6.1,II
Documents
Specifications
Specification Test Results
- Appearance:Clear yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5591 to 1.5611 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with HClO4:98.5 to 101.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



