4,4'-Dihydroxybenzophenone 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
4,4'-Dihydroxybenzophenone 97%
Catalog # ACRO114920250
Supplier: Thermo Fisher Scientific
CAS Number: 611-99-4
Jump to:
Product Details & Documents
Synonyms: Bis(4-hydroxyphenyl) ketone
- CAS Number:611-99-4
- Formula:C₁₃H₁₀O₃
- MDL Number:MFCD00002358
- Molecular Weight:214.22 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:217…222 °C
- Density:1,336 g/cm³ (20 °C)
- EINECS:210-288-1
Specifications
Specification Test Results
- Appearance:Off-white to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:217 to 222 °C
- GC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



