4,4'-Dihydroxybenzophenone 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4,4'-Dihydroxybenzophenone 97%
Catalog # ACRO114920050
Supplier: Thermo Fisher Scientific
CAS Number: 611-99-4
Jump to:
Product Details & Documents
Synonyms: Bis(4-hydroxyphenyl) ketone
- CAS Number:611-99-4
- Formula:C₁₃H₁₀O₃
- MDL Number:MFCD00002358
- Molecular Weight:214.22 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:217…222 °C
- Density:1,336 g/cm³ (20 °C)
- EINECS:210-288-1
Documents
ACRO11492
SDS
Specifications
Specification Test Results
- Appearance:Off-white to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:217 to 222 °C
- GC:≥96.0 %
- Titration with NaOH:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



