Cyclohexanone oxime 97%
Plastic bottle, 500 g
- :100-64-1
- :C₆H₁₁NO
- :MFCD00001660
- :113.16 g/mol
- :206 °C (1013 hPa)
- :88…92 °C
- :1,13 g/cm³ (20 °C)
- :90 °C
- :Ambient
- :202-874-0
- :1325
- Appearance:Off-white to beige to light brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- Titration with HClO4:≥96.0 %
- Pack type:Plastic bottle
- Pk:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



