Cyclohexanone oxime 97%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:100-64-1
- Formula:C₆H₁₁NO
- MDL Number:MFCD00001660
- Molecular Weight:113.16 g/mol
- Boiling Pt:206 °C (1013 hPa)
- Melting Pt:88…92 °C
- Density:1,13 g/cm³ (20 °C)
- Flash Pt:90 °C
- Storage Temperature:Ambient
- EINECS:202-874-0
- UN:1325
Specifications
Specification Test Results
- Appearance:Off-white to beige to light brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- Titration with HClO4:≥96.0 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g



