2-Iodophenol 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:533-58-4
- Formula:C₆H₅IO
- MDL Number:MFCD00013963
- Molecular Weight:220.01 g/mol
- Merck Index:13,05057
- Boiling Pt:186…187 °C (160 mmHg)
- Melting Pt:37…43 °C
- Density:1,8757 g/cm³ (80 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:208-569-9
- ADR:6.1,III
Documents
ACRO41248
SDS
Specifications
Specification Test Results
- Appearance:White to brown-red to brown Crystalline powder or low melting mass
- Infrared spectrum:Conforms
- Melting point:37 to 43 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



