Double spatulas, micro
210 mm, 55 mm, 6 mm, Green
Catalog # 231-2226
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel, PTFE coated.
Specifications
- Colour:Green
- Length:210 mm
- Blade width:6 mm
- Blade length:55 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



