Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol 98+%

3-Fluoropyrocatechol 98+%

Glass bottle for solids, 10 g
Catalog # ACRO264790100
3-Fluoropyrocatechol 98+%
Catalog # ACRO264790100
CAS Number:  363-52-0
3-Fluoropyrocatechol 98+%
3-Fluoropyrocatechol 98+%

Glass bottle for solids, 10 g

CAS Number:  363-52-0
Price
£132.00
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:71…74 °C
  • Density:1,514 g/cm³ (20 °C)

Documents

Specifications

Specification Test Results

  • Appearance:
    Light beige to grey-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    69 to 73 °C
  • GC:
    ≥98.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications