3-Fluoropyrocatechol 98+%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
3-Fluoropyrocatechol 98+%
Catalog # ACRO264790010
Supplier: Thermo Fisher Scientific
CAS Number: 363-52-0
Jump to:
Product Details & Documents
Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
- CAS Number:363-52-0
- Formula:C₆H₅FO₂
- MDL Number:MFCD00042582
- Molecular Weight:128.1 g/mol
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:71…74 °C
- Density:1,514 g/cm³ (20 °C)
Documents
ACRO26479
SDS
Specifications
Specification Test Results
- Appearance:Light beige to grey-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:69 to 73 °C
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



