Cycloheximide ≥90.0%
Glass bottle, 1 g
Inhibits DNA and protein synthesis.
: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- :66-81-9
- :C₁₅H₂₃NO₄
- :MFCD00082346
- :281.35 g/mol
- :Ambient
- :200-636-0
- :2811
- :6.1,I
Documents
Avantor Cell Culture Solutions.pdf
Product Literature
- Assay:Min. 90.0 %
- Melting point:107 - 120 °C
- Pack type:Glass bottle
- Pk:1 g



