Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Scalpels

155 - 160 mm, 35 mm, Broad blade
  233-0093
Scalpels
  233-0093
Scalpels
Scalpels

155 - 160 mm, 35 mm, Broad blade

 
£86.20
   
1 * 1 items
 

These scalpels are made from Wironit steel (18/12 stainless steel with 18% chrome and from 10 to 12% nickel).

  • Resistant to corrosion and anti-magnetic
  • With fixed blade and metal handle
 
  • Description:
    Broad blade
  • Length:
    155 - 160 mm
  • Seq:
    02
  • Blade:
    35 mm