Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Lead ACS

Lead ACS

Bottle
Catalog # APOSIN2182-100G
New Product
Supplier:   Apollo Scientific
Lead ACS
Catalog # APOSIN2182-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  7439-92-1
Lead ACS

Bottle

CAS Number:  7439-92-1
Price
£67.10
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Boiling Pt:1740 °C
  • Melting Pt:327.43 °C
  • Density:11.34 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications