Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cellulose D50:70~130, microcrystalline 90 μm

Cellulose D50:70~130, microcrystalline 90 μm

Bottle
Catalog # APOSBIC9004-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
Cellulose D50:70~130, microcrystalline 90 μm
Catalog # APOSBIC9004-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  9004-34-6
Cellulose D50:70~130, microcrystalline 90 μm

Bottle

CAS Number:  9004-34-6
Price
£198.00
Log in for availability
1 * 2,5 kg
Product Details & Documents

Low moisture type.

Usage: Biochemical/life science applications

Synonyms:  Microcrystallinecellulose
  • CAS Number:9004-34-6
  • MDL Number:MFCD00081512
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.