Cellulose D50:70~130, microcrystalline 90 μm
Bottle
Catalog # APOSBIC9004-100G
New Product
Supplier: Apollo Scientific
Cellulose D50:70~130, microcrystalline 90 μm
Catalog # APOSBIC9004-100G
Supplier: Apollo Scientific
New Product
CAS Number: 9004-34-6
Jump to:
Product Details & Documents
Low moisture type.
Usage: Biochemical/life science applications
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- MDL Number:MFCD00081512
Specifications
Specifications
- Pack type:Bottle



