4'-Hydroxyacetophenone
Bottle
Catalog # APOSOR11052-2.5KG
New Product
Supplier: Apollo Scientific
4'-Hydroxyacetophenone
Catalog # APOSOR11052-2.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 99-93-4
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:147…148 °C/3mmHg
- Melting Pt:108…110 °C
- Density:1.109 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:202-802-8
Specifications
Specifications
- Pack type:Bottle



