4'-Hydroxyacetophenone
Bottle
Catalog # APOSOR11052-25G
New Product
Supplier: Apollo Scientific
4'-Hydroxyacetophenone
Catalog # APOSOR11052-25G
Supplier: Apollo Scientific
New Product
CAS Number: 99-93-4
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:147…148 °C/3mmHg
- Melting Pt:108…110 °C
- Density:1.109 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:202-802-8
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



