Cyclohexanecarboxylic acid
Bottle
Catalog # APOSOR40331-25G
New Product
Supplier: Apollo Scientific
Cyclohexanecarboxylic acid
Catalog # APOSOR40331-25G
Supplier: Apollo Scientific
New Product
CAS Number: 98-89-5
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzoic acid
- CAS Number:98-89-5
- Formula:C₇H₁₂O₂
- MDL Number:MFCD00001461
- Molecular Weight:128.17 g/mol
- Boiling Pt:232…234 °C
- Melting Pt:27 °C
- Density:1.033 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



