2-Phenoxyethanol
Bottle
APOSOR4481-500G
New Product
: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- :122-99-6
- :C₆H₅OCH₂CH₂OH
- :MFCD00002857
- :138.17 g/mol
- :237…246 °C
- :41944 °C
- :1.107 g/cm³ (25 °C)
- :121 °C
- :Ambient
- :204-589-7
- Pack type:Bottle