4-Hydroxy-8-methoxyquinoline
Bottle
Catalog # APOSOR307979-100MG
New Product
Supplier: Apollo Scientific
4-Hydroxy-8-methoxyquinoline
Catalog # APOSOR307979-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 21269-34-1
Jump to:
Product Details & Documents
- CAS Number:21269-34-1
- Formula:C₁₀H₉NO₂
- MDL Number:MFCD02684163
- Molecular Weight:175.184 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



