Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Validation and Qualification Sets
Validation and Qualification Sets
Calibration set compatible with Classic line and Top line media preparators TP-VAL-MP, for adjustment of the equipment temperature probes to ensure accurate measurements against a known standard
Catalog # ESPITP-VAL-MP
New Product
Supplier:   RAYPA
Validation and Qualification Sets
Catalog # ESPITP-VAL-MP
Supplier:  RAYPA
New Product
Validation and Qualification Sets
Validation and Qualification Sets
Calibration set compatible with Classic line and Top line media preparators TP-VAL-MP, for adjustment of the equipment temperature probes to ensure accurate measurements against a known standard
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
They make it possible to obtain an independent reading of the culture medium temperature, separate from the one collected by the equipment’s flexible probe. Essential for the calibration and/or qualification of media preparators.
  • Recommended for distributors or service providers specialised in sterilisation equipment
    Ideal for customers with their own technical service team who will handle the qualification and maintenance throughout the equipment’s lifespan
In the validation process, it is crucial to have options that guarantee the proper calibration and qualification of the equipment. For this purpose, we offer two specific kits to address these needs: calibration set and qualification set.
Calibration set: Calibration of media preparators can be performed with the TP-VALMP calibration set, allowing for the adjustment of the equipment temperature probes to ensure accurate measurements against a known standard. This guarantees that the equipment operates within specified temperature limits.
Qualification set: To ensure that media preparators function according to design specifications, we offer three specialised qualification sets based on the selected model: TP-VAL-MP-20, TP-VAL-MP-40/60, and TP-VALMP-80/100. This process includes verification and documentation certifying that the equipment meets expected technical and performance standards. It is suitable for conducting both Installation Qualification (IQ) and Operational Qualification (OQ).
Ordering information: In order to use the calibration set, it is necessary to purchase the CAP-MP external probe adapter accessory.
Specifications
  • Description:
    Calibration set compatible with Classic line and Top line media preparators TP-VAL-MP, for adjustment of the equipment temperature probes to ensure accurate measurements against a known standard
  • Seq:
    1