Phthalide 99%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
Synonyms: Isobenzofuran-1(3H)-one
- CAS Number:87-41-2
- Formula:C₈H₆O₂
- MDL Number:MFCD00005906
- Molecular Weight:134,13203 g/mol
- EINECS:201-744-0
4 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Isobenzofuran-1(3H)-one
- CAS Number:87-41-2
- Formula:C₈H₆O₂
- MDL Number:MFCD00005906
- Molecular Weight:134,13203 g/mol
- EINECS:201-744-0
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



