Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Ammonium iodide 99%

Ammonium iodide 99%

Health
New Product
Supplier:   Apollo Scientific
Ammonium iodide 99%
Supplier:  Apollo Scientific
New Product
Certificates
Ammonium iodide 99%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:12027-06-4
  • Formula:NH₄I
  • Molecular Weight:144,94 g/mol
  • EINECS:234-717-7
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £16.50
    £16.50
    1 * 25 g
       £16.50

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £20.90
    £20.90
    1 * 100 g
       £20.90

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £96.80
    £96.80
    1 * 500 g
       £96.80

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - £475.00
    £475.00
    1 * 2,5 kg
       £475.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - £1,900.00
    £1,900.00
    1 * 10 kg
       £1,900.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.