Rucaparib Phosphate ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one phosphate
- CAS Number:459868-92-9
- Formula:C₁₉H₁₈FN₃O·H₃PO₄
- Molecular Weight:421,36 g/mol
3 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one phosphate
- CAS Number:459868-92-9
- Formula:C₁₉H₁₈FN₃O·H₃PO₄
- Molecular Weight:421,36 g/mol



