Software, SW7000
Communication software between the equipment and the PC, SW7000, Real-time display and recording, or post-cycle review
Catalog # ESPISW7000
Supplier: RAYPA
Jump to:
Product Details & Documents
Allows the visualisation, documentation and printing of the processes carried out in the AE-DRY, AES, AHS-DRY and AHS-N series autoclaves.
Includes RS-232 cable, USB stick with software, and RS-232 to USB adapter
- Real-time and post-cycle data analysis
Communication software between the equipment and the PC for display and recording in real time or display after each cycle. Cycles can also be printed or exported to excel. PC connection via RS-232. Data can also be exported directly to a USB memory stick.
Specifications
- Description:Communication software between the equipment and the PC, SW7000
- For:Real-time display and recording, or post-cycle review