Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
E7820 ≥98%

E7820 ≥98%

Marketsource
Supplier:   Abcam
E7820 ≥98%
Supplier:  Abcam
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

A small molecule sulfonamide derivative with potential antiangiogenic and antitumor activities. E7820 inhibits angiogenesis by suppressing integrin alpha 2, a cell adhesion molecule expressed on endothelial cells. Inhibition of integrin alpha 2 leads to an inhibition of cell-cell interactions, endothelial cell-matrix interactions, vascular endothelial cell proliferation and angiogenesis.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  3-Cyano-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulphonamide
  • CAS Number:289483-69-8
  • Molecular Weight:336,4 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 3-cyano-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulfonamide
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - £97.40
    £97.40
    1 * 5 mg
       £97.40

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - £339.00
    £339.00
    1 * 25 mg
       £339.00

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.