Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Pterostilbene ≥98% antiapoptotic agent antioxidant

Pterostilbene ≥98% antiapoptotic agent antioxidant

Catalog # ABCAAB120961-10
Marketsource
Supplier:   Abcam
Pterostilbene ≥98% antiapoptotic agent antioxidant
Catalog # ABCAAB120961-10
Supplier:  Abcam
Marketsource
CAS Number:  537-42-8
Pterostilbene ≥98% antiapoptotic agent antioxidant
CAS Number:  537-42-8
Price
£149.00
Log in for availability
1 * 10 mg
Product Details & Documents

Antioxidant and antiapoptotic agent; analog of resveratrol. Antineoplastic properties in vitroand in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Molecular Weight:256,3 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Pterostilbene