Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
BCECF AM ester ≥98% intracellular ratiometric pH-indicator

BCECF AM ester ≥98% intracellular ratiometric pH-indicator

Catalog # ABCAAB143463-1
Marketsource
Supplier:   Abcam
BCECF AM ester ≥98% intracellular ratiometric pH-indicator
Catalog # ABCAAB143463-1
Supplier:  Abcam
Marketsource
CAS Number:  117464-70-7
BCECF AM ester ≥98% intracellular ratiometric pH-indicator
CAS Number:  117464-70-7
Price
£144.00
Log in for availability
1 * 1 mg
Product Details & Documents

Acetoxymethyl ester of BCECF. Intracellular ratiometric pH indicator. Converts to fluorescent BCECF via the action of intracellular esterases. Ratio of fluorescence intensities can be converted to pH between 4 and 9. Excitation: 500 nm (high pH), 450 nm (low pH). Emission: 531 nm.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM
  • CAS Number:117464-70-7
  • Molecular Weight:556,48 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.