Adjustable Volume Pipettor, EX+
Catalog # COLE07911-21
Supplier: Cole-Parmer
New Product
Specifications
- Volume:100 - 1000 µl
- With:Tip ejector
- Description:Adjustable volume pipettor, non sterile
- Increment:2 µl
- Accuracy:±1.00 - ±0.70%
Specifications
About this item
Chemical-resistant PTFE O-rings and seals allows these pipettors to be used with solvents.
Fully autoclavable
UV resistant for flexible sterilisation
One-touch calibration
Enhanced ergonomics
Enhanced UV resistance provides added protection from biohazardous elements. Easily adjust the pipette volume by turning the coloured dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.
Body material: DS polyester urethane zzle filter; Al₂O₃ ceramic and PP plunger assembly