Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Human CD44 ELISA Set

Human CD44 ELISA Set

Human CD44 ELISA set, 100 tests
Catalog # ABCAAB212086-100
Marketsource
Supplier:   Abcam
Human CD44 ELISA Set
Catalog # ABCAAB212086-100
Supplier:  Abcam
Marketsource
CAS Number:  
Human CD44 ELISA Set
Human CD44 ELISA Set

Human CD44 ELISA set, 100 tests

CAS Number:  
Price
£882.00
Log in for availability
1 * 100 Tests
Product Details & Documents

Human CD44 ELISA Set is a Sandwich (quantitative) ELISA kit for the measurement of Human CD44 ELISA Set in Human in plasma, cell culture supernatant, serum samples.

Specifications

Specifications

  • Description:
    Human CD44 ELISA set
  • Tests per kit:
    100 tests
  • Cat. No.:
    ABCAAB212086-100
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,13 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Sample type:
    Plasma, Cell culture supernatant, Serum
  • Target protein:
    CD44
  • Detection range:
    0,31 - 10 ng/ml
  • Storage temperature:
    4 ℃
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.