Anti-beta Tubulin Mouse Monoclonal Antibody [clone: TUB 2.1] (Cy3®)
ABCAAB11309-50
: Abcam
- Seq:1202
- Size:50 µl
- Pack type:Vial
Cy3 ® Mouse monoclonal [TUB 2.1] to beta Tubulin.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: TUB 2.1
Conjugation: Cy3®
Epitope: Ab11309 specifically recognizes an epitope in the carboxy-terminal part of beta-tubulin (between amino acids 281-446)
Host: Mouse
Isotype: IgG1
Reactivity: Chicken