- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Total Immunoglobulin
- Description:Porcine Total Immunoglobulin ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Porcine Total Immunoglobulin ELISA Kit exhibits high specificity and excellent specificity for the detection of porcine Total Immunoglobulin. No significant cross-reactivity or interference between Total Immunoglobulin and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:1,875 - 120 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:1,125 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Porcine Total Immunoglobulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Total Immunoglobulin in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Porcine Total Immunoglobulin ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Total Immunoglobulin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Total Immunoglobulin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Total Immunoglobulin present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Total Immunoglobulin Antibody, which binds the captured Total Immunoglobulin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Total Immunoglobulin captured in each well. The concentration of Total Immunoglobulin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.