- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:ADCY5
- Description:Human ADCY5 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Human ADCY5 ELISA Kit exhibits high specificity and excellent specificity for the detection of human ADCY5. No significant cross-reactivity or interference between ADCY5 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,094 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Human ADCY5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ADCY5 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human ADCY5 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ADCY5 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ADCY5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ADCY5 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-ADCY5 Antibody, which binds the captured ADCY5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ADCY5 captured in each well. The concentration of ADCY5 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.