Human Cathepsin G Antibody ELISA Kit
Catalog # ANTIA319453-96
Supplier: ANTIBODIES.COM
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Cathepsin G Antibody
- Description:Human Cathepsin G Antibody ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Human Cathepsin G Antibody ELISA Kit exhibits high specificity and excellent specificity for the detection of human Cathepsin G Antibody. No significant cross-reactivity or interference between Cathepsin G Antibody and analogues was observed.
- Detection method:Colorimetric
- Time to Results:3 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:1,563 - 100 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,938 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human Cathepsin G Antibody ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Cathepsin G Antibody in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Delivery: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.
Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.