Order Entry
United Kingdom
ContactUsLinkComponent
 
Human Kininogen 1 ELISA Kit
Catalog # ANTIA314262-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Human Kininogen 1 ELISA Kit
Catalog # ANTIA314262-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Kininogen 1
  • Description:
    Human Kininogen 1 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    150 - 2400 nmol/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9,02 nmol/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human Kininogen 1 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Kininogen 1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay Precision: Intra-assay CV: 3,6%, Inter-assay CV: <10%

Human Kininogen 1 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Kininogen 1 in serum, plasma or other biological fluids. An antibody specific for Kininogen 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Kininogen 1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Kininogen 1 Antibody, which also binds the Kininogen 1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Kininogen 1 Antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Kininogen 1 Antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Kininogen 1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Kininogen 1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch and protocol manual.
Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.