SARS-CoV-2 Nucleoprotein ELISA Kit
ANTIA319530-96
New Product
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:SARS-CoV-2 Nucleoprotein
- Description:SARS-CoV-2 Nucleoprotein ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:SARS-CoV-2 Nucleoprotein ELISA Kit exhibits high specificity and excellent specificity for the detection of universal SARS-CoV-2 Nucleoprotein. No significant cross-reactivity or interference between SARS-CoV-2 Nucleoprotein and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 hours
- Shelf life:Store for 6 months at 4 °C
- Detection range:12,5 - 800 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:7,5 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
SARS-CoV-2 Nucleoprotein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of SARS-CoV-2 Nucleoprotein in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA kit
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
: This is a Ready-To-Use ELISA kit that contains: Pre-Coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.