Recombinant Human ErbB2/HER2 (from HEK293 cells)
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Petri dishes
Supplier: Anicrin
VWR®, Cryoboxes
Supplier: VWR Collection
Cryobank vials and Bank-It™ tube system, Nunc™
Supplier: Thermo Fisher Scientific
Cap inserts, PP, Corning®
Supplier: Corning
Filter plates, 24-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Microplates, 96-well
Supplier: Greiner Bio-One
Mass cell culture system, CELLdisc™
Supplier: Greiner Bio-One
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
Coloured cap inserts, Capinsert™
Supplier: Simport Scientific
Serological pipettes, Sterilin®
Supplier: STERILIN
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
R-50x8 Static strip processing assembly, research grade, 8 wells, 50 uL per well volume, pack of 3 1 * 3 items
Supplier: MARKET SOURCE PART PROCESS
Bottles, Square, PET
Supplier: LAB 3 BOTTLES
2 x 235 mL deep well plat
Supplier: MARKET SOURCE PART PROCESS
TILT bottles, DURAN®
Supplier: DWK Life Sciences