(S)-(-)-2-Methyl-CBS-oxazaborolidine 0.5 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, (S)-Me-CBS Catalyst, (S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
CAS Number:112022-81-8
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439
:277.17 g/mol
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together