

Bis(1,5-cyclooctadiene)nickel(0) ≥98%
Catalog # STRE28-0010-2G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2 g
- Seq:3107
Specifications
About this item
:
:275.06 g/mol
Melting Pt:60 °C (N2) (dec.)
EINECS:215-072-0
UN:1325
ADR:4.1,II
:GHS02
:GHS08
CAS Number:1295-35-8
Formula:C₁₆H₂₄Ni
MDL Number:MFCD00058902