Organic reference standard, Etidronic acid 10 µg/ml in methanol
Catalog # PROONE9256
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2809-21-4
- Description:Etidronic acid 10 µg/ml in methanol
- Formula:C₂H₈O₇P₂
- Description:Etidronic acid