Ethyl 1-naphthoate ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 3007-97-4 Molecular Formula: C13H12O2 Formula Weight: 200.24 Refractive Index: 1.5930 Boiling Point: 308-310° MDL No.: MFCD00016805
- CAS Number:3007-97-4
- Formula:C₁₃H₁₂O₂
- MDL Number:MFCD00016805
- Molecular Weight:200.24 g/mol
- Boiling Pt:308…310 °C
- Density:1.110
- Storage Temperature:Ambient
- EINECS:221-120-1
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 3007-97-4 Molecular Formula: C13H12O2 Formula Weight: 200.24 Refractive Index: 1.5930 Boiling Point: 308-310° MDL No.: MFCD00016805
- CAS Number:3007-97-4
- Formula:C₁₃H₁₂O₂
- MDL Number:MFCD00016805
- Molecular Weight:200.24 g/mol
- Boiling Pt:308…310 °C
- Density:1.110
- Storage Temperature:Ambient
- EINECS:221-120-1
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



