Order Entry
United Kingdom
ContactUsLinkComponent
Progesterone
Progesterone
Catalog # SERA33530.03
Supplier:  Serva
CAS Number:  
Progesterone
Catalog # SERA33530.03
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
EINECS:200-350-6
CAS Number:57-83-0
Formula:C₂₁H₃₀O₂
MDL Number:MFCD00003658