Order Entry
United Kingdom
ContactUsLinkComponent
Tri-tert-butylphosphonium tetrafluoroborate ≥99%
Irritant
Tri-tert-butylphosphonium tetrafluoroborate ≥99%
Catalog # STRE15-6000-1G
CAS Number:  
Irritant
Tri-tert-butylphosphonium tetrafluoroborate ≥99%
Catalog # STRE15-6000-1G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.13 g/mol
Melting Pt:261 °C (dec.)
 :GHS07
CAS Number:131274-22-1
 :MFCD07189501
Formula:C₁₂H₂₈BF₄P
MDL Number:MFCD04039975