n-Butyl phenyl ether, Sigma-Aldrich®
Catalog # SIAL142344-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.22 g/mol
:M-Clarity™ quality level = MQ100
EINECS:214-426-1
CAS Number:1126-79-0
Formula:C₁₀H₁₄O
MDL Number:MFCD00009438