D(+)-α-Acetobromoglucose 98.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose
CAS Number:572-09-8
:
Formula:C₁₄H₁₉BrO₉
MDL Number:MFCD00063254
Molecular Weight:411.2 g/mol
Melting Pt:83…88 °C
EINECS:209-339-0
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to beige to light grey Powder
- Infrared spectrum:Conforms
- Melting point:84 to 89 °C
- Titration Argentometric:≥98.0 %
- Specific optical rotation:+188° to +202° (20 °C, 589 nm) (c=2, CHCl3)
Specification Test Results
Frequently Bought Together